Rubbermaid® Commercial TradeMaster® Utility Carts
Supplier: Janitorial Supplies
RCP453388BLA
19758-924EA 5940.4 USD
19758-924
Rubbermaid® Commercial TradeMaster® Utility Carts
Carts
The Rubbermaid® Commercial TradeMaster® utility cart easily transport tools and supplies where you need them.
  • Includes shelf, drawers, and cabinet
  • Transports up to 750 lbs
  • Structural foam body for high strength and low weight
The utility cart is made from a durable structural foam construction. The rolling utility cart is low maintenance, and rust, dent, and chip resistant. These utility carts feature a comfort grip, user-friendly handle and molded-in compartments for small parts or tools. The utility cart includes 5' non-marking casters, 2 fixed and 2 swivel with total lock brakes and full-extension, ball bearing drawer slides. Single key lock secures all drawers and allows one drawer to open at a time.
Ordering information: Available accessories (sold separately) include a 56 cm (22") refuse bin that holds 30 l (8 gal.), a 26.7 cm (101/2") utility bin that holds 15 l (4 gal.); a molded parts organizer that fits on top shelf of cart, a three-outlet surge protected power strip with 4.6 m (15') cord, a set of four hooks for holding cables and step stools; and a 20.3 cm (8") pneumatic caster kit that allows smoother operation on rough surfaces. Contact your VWR representative for more information.