Heavy-Duty Utility Carts, Lakeside
Supplier: Lakeside
721 722 743 758 954 953 944 943 744
19758-080EA 2474.37 USD
19758-080 19758-082 19758-086 19758-088 19758-344 19758-346 19758-348 19758-350 19758-352
Heavy-Duty Utility Carts, Lakeside
Carts
Designed for heavy duty industrial loads, carts are 11-gauge stainless steel, angle constructed and feature 20.3cm (8") rigid no-mark polyurethane casters on the rear with 12.7cm (5") no-mark polyurethane swivels on the front, which provide great rollability under heavy loads
Note: Capacity is the total cart capacity across all of the shelves with the weight evenly dispersed.
Shelves have a 2.5cm (1") lip on three sides. Carts are shipped fully assembled.