Stereo Microscopes, K Series, Motic
Supplier: Motic
19000-218 19000-268 19000-222 19000-224 19000-250 19000-296 19000-298 19000-204 19000-208 19000-210 19000-216 19000-228 19000-252 19000-254 19000-286 19000-272 1101001701642 19000-280 19000-282 19000-284 19000-288
19000-218EA 1896.13 USD
19000-218 19000-268 19000-222 19000-224 19000-250 19000-296 19000-298 19000-204 19000-208 19000-210 19000-216 19000-228 19000-252 19000-254 19000-278 19000-286 19000-270 19000-272 19000-274 19000-276 19000-280 19000-282 19000-284 19000-288
Stereo Microscopes, K Series, Motic
Microscopes
These stereo microscopes feature a pair of 10x, 23mm super widefield eyepieces and a 45° inclined binocular tube with dioptric compensation on both tubes and interpupillary distance adjustment between 54mm and 76mm
Choose from the K-400 series microscopes with a four-step magnification changer at 6x, 12x, 25x, and 50x ratios; the K-500 series microsopes with a five-step magnification changer at 6.4x, 10x, 16x, 25x, and 40x ratios; or the K-700 series microscopes that feature a zoom range at 1:5.2 ratio designed on self-centering dovetail ring mechanism, total parfocality, and absolute centering.
Microscopes come in either a streamlined stand with a large working surface or a light stand with built-in incident and transmitted 12V, 10W halogen light with intensity control and enhancing choices of top, bottom, or top and bottom light. Microscopes with the built-in halogen light feature dual voltage with a 110V main supply or 220V.
All microscopes come with a standard 1.0x objective.
Ordering information: Heads with focusing mounts, eyepieces, objectives, and other accessories sold separately.