Stak-a-Cab™ Safety Storage Cabinets, SciMatCo
Supplier: Scientific Materials
SC9042 SC2260 SC1360 SC2560 SC1460 SC2460 SC1560
14212-946EA 1806.22 USD
14212-946 14212-948 14212-954 14212-958 18500-050 18500-054 18500-060 18500-062 14212-960 18500-056 18500-058
Stak-a-Cab™ Safety Storage Cabinets, SciMatCo
Furniture Storage Cabinets
Choose from a variety of specialized stackable cabinets for the storage needs.
  • Cabinets are easily identified by cabinet color and (8 to 9") hazard label
  • Mini Stak-a-Cab™ cabinets offer the same level of protection in smaller units
Available as an accessory, Polypropylene isolation compartment (18500-072) fits in Stak-a-Cab™ units and isolates nitric acid or nitric acid spills from all other products stored in these cabinets.
All cabinets are constructed of 2.5cm (1") thick multi-ply plywood and finished with clear epoxy paint. Individual cabinets are designed for stacking or as stand-alone units for the laboratory bench and allow for the safe organization of compatible chemicals. All Stak-a-Cabs include polypropylene liners on the interior bottom of the cabinet for easy clean-up of accidental spills.
Certifications: All cabinets are supplied with suitable locking mechanisms and are constructed to meet all applicable NFPA and OSHA standards.