Ranger Surface-Mix Hand Torch, Wale Apparatus
Supplier: Wale Apparatus
90-9300
18363-005EA 826.63 USD
18363-005
Ranger Surface-Mix Hand Torch, Wale Apparatus
Heaters Gas Burners
Provides a hot quiet flame without flashback
Lightweight stainless steel construction with wood handle and nickel-plated brass valve. Performs well with fuel pressures from 0.25 to 5 psi for natural gas and propane; from 0.25 to 15 psi for oxygen.
Ordering information: Order torch and twin-tubing connectors separately.