Cole-Parmer® BB-200 Series Electric Bunsen Burners, Antylia Scientific
Supplier: Argos Technologies
36135-04
18344-000EA 1108.76 USD
18344-000
Cole-Parmer® BB-200 Series Electric Bunsen Burners, Antylia Scientific
Heaters Electric Burners
Provides clean and easy operation.
  • Top cowl deflects heat away from hands
  • Air circulation from vented housing keeps base cool to touch
  • Energy efficient - only consumes 400 W of power
This electric Bunsen burner combines the advantages of a regular gas burner with the clean and easy operation of a heating mantle. Corrosion-resistant stainless steel burner features a cone-shaped coiled resistance heater which directs radiant heat out from the cavity. Cooling vents circulate air to cool the base, making it possible to hold while in use. A detachable cowl also deflects heat from the medium surface.
Ideal for non-contact radiant heating of materials in test tubes, crucibles, small flasks, and beakers. Effective heating of a test tube sample should be done 25 mm above the top of the burner. Crucible heating may be achieved by removing the cowl.
Note: To regulate the power input and temperature of the Bunsen burner, a heating controller is required. Without a controller, the burner operates continuously at full heating power. The Bunsen burner can be purchased with or without the recommended controller.
Ordering information: Supplied with power cord with US plug.