SafeCare™ Fluid-Resistant, Antistatic, Reusable Lab Coats, Encompass
Supplier: Encompass
46942-115 46942-117 46942-119 46942-112XL
17788-044EA 117.42 USD
17788-044 17788-046 17788-048 17788-050
SafeCare™ Fluid-Resistant, Antistatic, Reusable Lab Coats, Encompass
Workwear
Designed as a substitute for traditional lab coats when extra fluid protection is required.
  • Coats feature collar-closure snap front, breast pocket, and two patch pockets
  • Lapel collar and knit elastic cuff
  • Manufactured from 99% polyester/1% carbon fiber
  • Available in white or blue
The use of carbon fibers helps dissipate harmful static. Fabric will maintain its fluid repellency up to 100 washings. Coats are non-linting and stain-resistant.