PYREX® Dispensing Burets, Colored Scale, with Straight Bore PTFE Stopcock, Corning
Supplier: Corning
2094-250 2094-500 2094-1L
17475-207CS 315.05 USD
17475-207 17475-229 17475-241
PYREX® Dispensing Burets, Colored Scale, with Straight Bore PTFE Stopcock, Corning
Burets
Burets are mainly designed to dispense large volumes of liquids rapidly and accurately. Suitable for use in school and institutional laboratories not requiring the accuracy of Class A models.
  • Made from close tolerance, accurate bore tubing with uniform walls
  • Tips carefully drawn and ground to reduce chipping
  • With permanent coloured scale
Provided with a PTFE stopcock plug to reduce freezing and eliminate lubricant contamination. The smooth micro-finish of the stopcock barrel assures a precision fit and optimum leak-resistant performance.