PYREX® Burets, Class A, Colored Graduations, Fluoropolymer Resin Stopcock, Corning
Supplier: Corning
2103-10 2103-50 2103-100 2103-25
17452-368CS 274.09 USD
17452-368 17452-404 17452-426 17452-380
PYREX® Burets, Class A, Colored Graduations, Fluoropolymer Resin Stopcock, Corning
Burets
Borosilicate glass, class A.
  • Red colored graduations feature permanent markings, fine clear lines
  • Large easy to read numbers
  • Smooth micro-finish stopcock assures leak-resistant performance
Burette tips are carefully drawn from precision bore tubing to ensure proper drainage rates.
Certifications: Capacity tolerance is established by ASTM E287 and calibrated in accordance with ASTM E542.
Delivery information: Burets are shipped complete with PTFE stopcock and dust cover.