Wire Brush, Mill-Rose
Supplier: Mill-Rose
MR594801
17263-004EA 42.37 USD
17263-004
Wire Brush, Mill-Rose
Brushes
Fine-gauge, brass wire brush can be used to clean dirty metal surfaces or prepare cement molds for new samples.
Curved hardwood handle provides sure grip with good hand clearance.
Wire size: 0.15 mm (0.006")