Cylinder Brush, Justman
Supplier: Justman Brush
J720238
17252-120EA 59.39 USD
17252-120
Cylinder Brush, Justman
Brushes
Full radial tip provides thorough cleaning action and prevents the brush from scratching glassware.
Ideal for washing bottles, cylinders, and large test tubes.