SP Bel-Art Digi-Max™ Slide Caliper with LCD Readout, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H13417-0000
17150-058EA 444.33 USD
17150-058
SP Bel-Art Digi-Max™ Slide Caliper with LCD Readout, Bel-Art Products, a part of SP
Calipers
Take measurements with speed and ease using the Digi-Max™ digital slide caliper.
  • Easy-to-read LCD display
  • Long-life battery
  • Non-conducting
  • Corrosion-resistant
  • Lighweight
The LCD display immediately indicates the exact measurement taken with push button selection between metric and English scales and a self-zeroing feature for accuracy of ± 0 .1 mm (0.004")
A long-life battery is included and is easily accessible for replacement. On/Off button preserves battery life. Molded-in rigid plastic, they will not scratch like metal calipers can. Supplied in a protective plastic case.
Metric scale to 150 mm and English scale to 6". These calipers are non-conducting, corrosion resistant, and lightweight for easy handling.