Babcock Cream Test Bottle Brush, Mill-Rose
Supplier: Mill-Rose
72220B
17065-000PK 43.99 USD
17065-000
Babcock Cream Test Bottle Brush, Mill-Rose
Brushes
Special construction puts the cleaning action directly where it is needed.
  • Along the brush stem, rows of short dense bristles provide extra scrubbing for the neck
  • All natural black bristles twisted into a wire handle with a hanging loop
The extra long radial bristles fold to fit through the narrow bottle neck then expand to fill the chamber.
Neck diameter: 1.3 cm (1/2")