Babcock Milk Test Bottle Brush, Mill-Rose
Supplier: Mill-Rose
72223W
17050-003PK 41.52 USD
17050-003
Babcock Milk Test Bottle Brush, Mill-Rose
Brushes
Designed specifically for cleaning the narrow necks of Babcock milk test bottles.
  • Short dense bristles reach easily into tall-form bottle necks
  • Natural white bristles twisted into a wire handle with hanging loop