Microcentrifuge Tube Brush, Tufted End, Mill-Rose
Supplier: Mill-Rose
72201W
17030-006PK 37.22 USD
17030-006
Microcentrifuge Tube Brush, Tufted End, Mill-Rose
Brushes
Narrow nylon brush with tapered end suitable for conical and round bottom tubes.
Short dense bristles held in a twisted wire handle with hanging loop.
Brush taper: 0.6–1.3 cm (1/41/2")