PIE-Blocks, Reaction Blocks, Chemglass
Supplier: Chemglass
CG-1991-P-01
17006-592EA 975.23 USD
17006-592
PIE-Blocks, Reaction Blocks, Chemglass
Block Heaters Blocks
Reaction PIE-BLOCK is designed to fit directly on 135 mm diameter circular top hot plate stirrers
  • Holes are spaced for optimum stirring of all vials
  • Inserts are color coded for quick and easy identification
  • Interchangeable pie wedges permit various sized vials on one hot plate
  • Aluminum construction provides excellent heat transfer
  • Safety pie plate holder keeps up to four pie wedges securely locked together on hotplate
A drilled thermowell is provided for insertion of contact thermometers or temperature sensors.
Ordering information: Supplied with safety Holder and 6 listed Pie Wedges.