Sodium pyruvate solution 100 mM, HyClone™

Supplier: Cytiva

SH30239.01
16777-188EA 14.98 USD
16777-188
Sodium pyruvate solution 100 mM, HyClone™
Sodium pyruvate

Sodium pyruvate solution is an easily accessible carbohydrate source that may also have protective effects against hydrogen peroxide.


Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.

Formula: CH₃COCOONa
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

Specification Test Results

Endotoxin concentration ≤10.00 EU/mL
Sterilization 0.1 µm sterile filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR