Penicillin : Streptomycin solution, HyClone™
Supplier: Cytiva
SV30010
16777-164EA 30.06 USD
16777-164
Penicillin : Streptomycin solution, HyClone™
Penicillin/Streptomycin
HyClone™ Penicillin-Streptomycin Solution (100X) is a broad spectrum antibiotic designed to control and destroy destructive microorganisms, such as gram negative and gram positive bacteria.
  • Used as an antibacterial or antimycobacterial
  • Penicillin interferes with the final stage of synthesis of the bacterial cell wall
  • Streptomycin, through protein synthesis, inhibits elongation at transpeptidation step; binds to 30S subunit causing misreading
Common working concentration for Penicillin is 100 U/ml and for Streptomycin is 100 μg/ml.
CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)