HyClone™ HEPES Buffer Solution 1 M, HyClone products (Cytiva)
Supplier: Cytiva
Maintain physiological pH in cell culture, despite changes in carbon dioxide concentration (produced by cellular respiration) with Cytiva HyClone HEPES solution.
- 0.1 µm sterile filtered
- pH: 7.2 - 7.5
- Storage: 2 to 8° C
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...