Stackable Cylindrical Parr Weighing Bottle, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
15157-2024
16705-006CS 577.04 USD
16705-006
Stackable Cylindrical Parr Weighing Bottle, Kimble®, DWK Life Sciences
Bottles Weighing Bottles
KIMAX® bottle designed for weighing small samples of coal.
  • Capacity to neck base: 4 ml
  • Cap-style closure fits over externally ground body
  • 15182 cap-style stopper may be used as a replacement
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.