VWR® Wash Bottle Carrier
Supplier: VWR International
16649-980
16649-980EA 70.72 USD
16649-980
VWR® Wash Bottle Carrier
Carriers Bottle Carriers/Containers
Sturdy, lightweight polypropylene carrier can hold up to six wash bottles.
  • Polypropylene construction
  • Sturdy
  • Lightweight
  • Secure, comfortable handles
Its secure handle provides comfort and control. Carrier is chemical-resistant. It holds both 237 mL (8 oz.) and 473 mL (16 oz.) bottles. Autoclavable at 121°C (250°F).