Dangerous Goods UN EPS Foam Packaging, Qorpak
Supplier: Berlin Packaging
HMS-6551M
16465-066CS 343.83 USD
16465-066
Dangerous Goods UN EPS Foam Packaging, Qorpak
Mailers and Shippers
EPS Foam provides better protection and cushioning than standard fibreboard packaging.
Certifications: Meets UN 4G standards for Packing Groups II and III.
Ordering information: This package contains a pressure tested amber plastic coated glass jug and phenolic cap. For air shipments, check the regulations for any absorbent requirements and quantity limitations. Includes LDPE bag. Absorbent is sold separately.
Packaging: Sold in a convenience pack of 4.