Dangerous Goods DOT-SP 9168 UN 4GV Packaging, Qorpak
Supplier: Berlin Packaging
HMS-6649M
16465-052CS 647.96 USD
16465-052
Dangerous Goods DOT-SP 9168 UN 4GV Packaging, Qorpak
Mailers and Shippers
Utilize our Special Permit, DOT-SP 9168, to ship approved hazardous materials without labeling, placarding, or segregation domestically, or meet the 4GV requirements for international transport.
  • DOT-SP 9168 provides relief for labeling, placarding, and segragating of select materials
  • Primary Container meets 95 kPa requirements for air transport
This package is required by many small parcel carriers for the transport of select dangerous goods domestically. Those same carriers might also require 4GV packaging for international shipments. Feel at ease with a pressure tested primary container and absorbent material (both provided).
Certifications: Meets UN 4GV standards for Packing Groups I, II, and III, as well as carrying the DOT-SP 9168 Special Permit.
Delivery information: Pressure tested wide mouth plastic coated glass bottle, phenolic F217/PTFE lined cap, and absorbent pouch included.
Packaging: Sold in a convenience pack of 4.