Steel Closed Head Container, UN Marked, Qorpak®
Supplier: Berlin Packaging
HMS-6741M
16465-019CS 472.79 USD
16465-019
Steel Closed Head Container, UN Marked, Qorpak®
Mailers and Shippers
One-gallon steel closed head drum is suspended in a die-cut carrier.
  • Unlined interior
  • Complete with plastic bag, tape and instructions
  • Pressure capability of 250 kPa
Acceptable for Great Danger Hazardous Materials (gross mass not exceeding 9.5 kg) Packing Group I.