SP Bel-Art Gas Washing Bottle with Fritware® Dispersion Disc, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H11030-0000
16404-701CS 492.26 USD
16404-701
SP Bel-Art Gas Washing Bottle with Fritware® Dispersion Disc, Bel-Art Products, a part of SP
Bottles Gas Washing Bottles
Graduated 250 ml polypropylene cylinder, with a stable extended base for use in precipitating elements from gas.
  • Practical and ecnomical
  • Coarse grade porosity disc
Gas is dispersed through a removable fitting that has a coarse grade porosity, polyethylene dispersion disc. The silicone stopper holds 6.3 mm (¹/₄") Ext.Ø tubes for entrance and exit of gas. Maximum pressure is 0.5 psi.