PET Bottle, Transparent, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
221135
16300-024CS 289.42 USD
16300-024
PET Bottle, Transparent, Wheaton®, DWK Life Sciences
Bottles Roller Bottles
Wheaton PET Bottles offer the clarity of glass with the safety of plastic.
  • Natural color - clear and transparent
  • Good alcohol and solvent barrier
  • Good gas and fair moisture barrier
  • Good to fair chemical barrier; not good for strong acids or bases
  • Good moldability
  • Sterilizable through EtO and gamma radiation
Polyethylene Terephthalate (PET) Characteristics: Semi-rigid to rigid depending on wall thickness. Good stress crack and impact resistance at room temperature and above.
The 120mL bottle is cylinder round style bottle and the smaller sizes have sloped shoulders. The narrow mouth is ideal for liquids. The bottles come with pre-attached white polypropylene screw cap with foamed polyethylene liner.