Nalgene® Rapid-Flow™ Filter Units and Bottle Top Filters, PES Membrane, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
165-0045 166-0045 167-0045 169-0045 295-3345 296-3345 296-4545 565-0020 595-3320 596-3320 597-3320 597-4520 565-0010 568-0010 566-0010 168-0045 295-4545 566-0020 567-0020 568-0020 569-0020 595-4520 596-4520 564-0020 567-0010
16211-053CS 137.02 USD
16211-053 16211-054 16211-056 16211-064 16211-066 16211-073 16211-077 73520-982 73520-992 73520-996 73521-000 73521-002 97014-830 97014-832 97014-834 16211-061 16211-068 73520-984 73520-986 73520-988 73520-990 73520-994 73520-998 82030-938 89176-982
Nalgene® Rapid-Flow™ Filter Units and Bottle Top Filters, PES Membrane, Sterile, Thermo Scientific
Filters Bottle Top Filters
All of these filters come with an asymmetric polyethersulfone (PES) membranes.
  • Graduated upper chambers hold from 150 to 1000 ml
  • Asymmetric PES membrane is an extremely clean, low-extractable, fast-flowing membrane
  • Provides excellent throughput and low protein binding of media and other solutions
  • 0.2 µm pore size membrane provides a last line of defense against bacterial contamination
  • 0.1 µm pore size membranes also protect against mycoplasma contamination
  • 0.45 µm pore size membranes are suitable for particle analysis, scintillation counting, prefiltration
More Product Information