Nalgene® Autoclavable Septum Closure, PP closure, Thermoplastic Elastomer Septum, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS2168-0384
16210-081CS 92.03 USD
16210-081
Nalgene® Autoclavable Septum Closure, PP closure, Thermoplastic Elastomer Septum, Thermo Scientific
Bottles Cell Culture Bottles
Thermo scientific nalgene autoclavable septum closures allow aseptic injection of reagent or sample withdrawal without compromising sterility or integrity of contents.
Suitable for use with any bottle or container with 38-430 neck, including Nalgene culture vessels.