Nalgene® Multi-Bottle Rack; White PC, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS5996-0871
16161-850EA 131.14 USD
16161-850
Nalgene® Multi-Bottle Rack; White PC, Thermo Scientific
Carriers Bottle Carriers/Containers
Nalgene White Polycarbonate Multi-Bottle Rack holds up to eight 500mL round or square bottles securely
The bottles fit snugly between upper grid and molded-in depression on lower tray. The multi racks contain handle telescopes for convenient storage.