Safety-Coated Media Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
219455 219457
16149-300CS 604.99 USD
16149-300 16149-302
Safety-Coated Media Bottles, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
USP Type I clear borosilicate glass.
  • Plastisol coating for safety
  • Provided with graduations and writing patch
  • Autoclavable