Nalgene® Carboys with Handles, Wide Mouth, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2235-0020 2235-0050
16133-502CS 929.83 USD
16133-502 16133-546
Nalgene® Carboys with Handles, Wide Mouth, Polypropylene, Thermo Scientific
Bottles Carboys
Ideal for microbiological materials, especially powder and solid sample handling.
  • Autoclavable
  • Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Note: Autoclave using a properly vented closure.
Wide mouth design allows for easy, efficient cleaning. Excellent chemical resistance. Graduated to contain in 1 gal. and 5 L increments. Cap size: 100-415.