Nalgene® Fluorinated High-Density Polyethylene, Wide-Mouth Large Bottle, Thermo Scientific
Supplier: Thermo Fisher Scientific
2124-0005
16130-015CS 274.56 USD
16130-015
Nalgene® Fluorinated High-Density Polyethylene, Wide-Mouth Large Bottle, Thermo Scientific
Bottles Plastic Bottles
The Nalgene® Fluorinated High-Density Polyethylene Wide-Mouth Large Bottle features a fluorinated surface that provides improved barrier properties and reduces solvent absorption and permeation.
  • Safe, durable, cost-efficient line and the fluorination enhances long-term container performance
  • Prevents material loss due to permeation and yields lower extractables
  • Wide-mouth design allows for easy filling and dispensing of solid and liquid materials
  • Bottles are split- and puncture-resistant, as well as recyclable