Nalgene® Large Square Bottle, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2123-0010
16130-010CS 251.52 USD
16130-010
Nalgene® Large Square Bottle, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Excellent for collecting, storing, and transporting dry chemicals, environmental samples, reagents, and distilled water.
Useful for low temperature applications. Molded-in handgrips provide greater safety and convenience when pouring. Wide mouth design allows easy access to contents. Leakproof. Cap size: 100-415.
Ordering information: Supplied with polypropylene screw caps assembled on bottles.