VWR® Standard Standard Stem Funnel
Supplier: VWR International
16126-904
16126-904CS 527.85 USD
16126-904
VWR® Standard Standard Stem Funnel
Funnels Filtration Funnels Filter Funnels
Polypropylene funnels feature an exclusive filter-step internal design for speedier filtrations.
  • External ribs prevent airlock
  • Parallel stem measures 8mm O.D.
  • Steam autoclavable at 121°C (250°F)
External ribbing prevents air lock. Resistant to most chemicals.