SP Bel-Art Dry-Keeper™ Small, Stacking Gas-Purge Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H42053-0002
16126-666EA 475.93 USD
16126-666 490012-530
SP Bel-Art Dry-Keeper™ Small, Stacking Gas-Purge Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccators, Plastic
Small, clear polystyrene desiccator cabinet safely stacks up to 5 units high for maximum use of lab bench space.
  • Polystyrene construction
  • Door disappears beneath cabinet when open
  • Clear siding
  • Stackable design
The pull-down door slides out of the way into a compartment beneath the cabinet.
Rear-mounted gas ports are tapered to accept tubing with 1/4 to 3/8" I.D. and can be closed to use desiccator with certian standard desiccant cartridges; Contact your Avantor representative for more information.