Nalgene® Square Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2018-0030 2018-0060 2018-0125 2018-0250 2018-1000
16120-701CS 162.24 USD
16120-701 16120-708 16120-715 16120-719 16120-728
Nalgene® Square Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Designed for long-term storage of chemicals and specimens where sterility is not required.
  • Leakproof
  • Not autoclavable
Bottles provide excellent chemical resistance, and withstand temperatures from –100 to 120 °C (–148 to 248 °F). The square design is easy to handle, and saves space on benchtop or shelf, in refrigerators and freezers, and under the lab hood. Permanent graduations on all sizes.
Ordering information: Supplied with polypropylene screw caps assembled on bottles. For replacement caps for 16120-715 through -728, see 16219-122 series.