Nalgene® Lowboy™ Carboys with Spigot, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2323-0008
16117-969CS 908.66 USD
16117-969
Nalgene® Lowboy™ Carboys with Spigot, High-Density Polyethylene, Thermo Scientific
Bottles Carboys
Ideal for storing and dispensing reagents, titrants, and distilled water.
  • Leakproof
For replacement caps, see cap 16219-104 in 16219-030 series.
Tolerates strong acids and bases. Not recommended for use with aggressive organic solvents. Low-profile design fits easily in tight spaces, on shelves, or in refrigerators. Can be stacked three high. Convenient integral handgrip for safe, easy transport. Cap Size: 63-415.
Ordering information: Supplied with carboy, polypropylene spigot, and screw cap.