Nalgene® Carboys with Handles, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
2250-0130 2250-0020 2250-0050
16101-142EA 539.6 USD
16101-142 16101-084 16101-109
Nalgene® Carboys with Handles, Polypropylene, Thermo Scientific
Bottles Carboys
Excellent for storing culture media, distilled water, and other solutions.
  • Leakproof
  • Autoclavable
For replacement caps, see cap 16225-234 in 16219-122 series.
Note: Autoclave using properly vented closures.
Graduated to contain in 1 gal. and 5 L increments. Carboys are equipped with two wide shoulder handles. Cap size: 83 mm. Accepts No. 13¹/₂ rubber stoppers.