Bottles, Cylinder, White High-Density Polyethylene, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLA-05751
16087-404CS 164.2 USD
16087-404 16087-408 89204-908
Bottles, Cylinder, White High-Density Polyethylene, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
White HDPE cylinder bottles are impact resistant.
They provide an excellent moisture barrier and offer a degree of light protection. Ideal for pharmaceutical applications. Narrow mouth cylinders are perfect for handling liquid samples in the lab. The shoulder style allows you to pour small quantities without spilling. HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping.