Nalgene® Carboy Vent Filter, Thermo Scientific
Supplier: Thermo Fisher Scientific
223-0030
16060-120CS 457.9 USD
16060-120
Nalgene® Carboy Vent Filter, Thermo Scientific
Filters Venting Filters
For venting and solvent filtration applications and for maintaining purified water stored in carboys.
  • Non sterile
  • Autoclavable
Filter can be used up to five times in venting applications. Stepped barbed hose fitting accommodates 6.4 to 12.7 mm (¹/₄ - ¹/₂") Int.Ø tubing.
The disposable unit features a Teflon® PTFE membrane with polypropylene housing. Membrane permits sterile venting on a slow exhaust/liquid autoclave cycle of NALGENE polypropylene and polycarbonate carboys up to 50 L.