Rosetta™ 2 Competent Cell Set, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
71405-3
16003-950EA 352.21 USD
16003-950
Rosetta™ 2 Competent Cell Set, Expression Strains, MilliporeSigma
Competent Cells
Rosetta™ 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
  • Supply tRNAs for 7 rare codones (AGA, AGG, AUA, CUA, GGA, CCC, and CGG)
  • Compatible chloramphenicol-resistant plasmid
  • tRNA genes are driven by their native promoters
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Delivery information: It includes Rosetta 2 Competent Cells (2×0.2 ml); Rosetta 2(DE3) Competent Cells (2×0.2 ml); Rosetta 2(DE3)pLysS Competent Cells (2×0.2 ml); 4×2 ml SOC Medium and 2 ng Test Plasmid.