Reaction Vessel Lid, 5-Necks, 150 mm, Chemglass
Supplier: Chemglass
CG-1946-40
16002-862EA 1385.42 USD
16002-862
Reaction Vessel Lid, 5-Necks, 150 mm, Chemglass
Lids Reaction Vessel Lids
Reaction vessel lids feature five-necks.
  • Five neck reaction vessel lid
  • For 150 mm flanged vessels
Lid has a 29/42 outer CN, two 34/45 outer SN’s, one Vertical 45/50 outer SN and one 45/50 outer SN angled 20 ° for powder addition.