Offset Adapters, Chemglass
Supplier: Chemglass
CG-1033-45 CG-1033-01 CG-1033-02
16002-606EA 234.89 USD
16002-606 80065-940 80065-942
Offset Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
The offset of the adapters is measured from center to center.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • Both joints are same size
The top outer and lower inner [ST] joints are equivalent.