Hematoxylin, dry powder stain

Supplier: Epredia

90022
16002-180EA 199.02 USD
16002-180
Hematoxylin, dry powder stain
Hematoxylin

Stain histology and cytology specimens progresively with Epredia™ Dry Powder Stains, certified by the Biological Stain Commission.

Formula: C₁₆H₁₄O₆
MDL Number: MFCD00078111
CAS Number: 517-28-2
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR