Eosin Y (yellowish), dry powder stain

Supplier: Epredia
Warning

Synonyms: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

90017
16002-170EA 334.52 USD
16002-170
Eosin Y (yellowish), dry powder stain
Eosin Y (yellowish)

Stain specimens with Eosin-Y compounds with Epredia™ Dry Powder Stains, certified by the Biological Stain Commission.


Enhances Differentiation Between Muscle, Red Blood Cells, and Connective Tissue

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR