VWR® Sample Container with Pour Spout
Supplier: VWR International
15704-050
15704-050CS 1237.02 USD
15704-050
VWR® Sample Container with Pour Spout
Sample Containers General Purpose Containers
Clear, graduated polystyrene container features a pour spout and a paper lid.
  • Pour spout
  • Clear siding
  • Graduated markings
  • Polystyrene construction
Graduations are marked from 10 to 200cc in 10cc intervals and from 1/2 to 61/2oz. in 1/2 oz. intervals. Height: 9.1 cm (39/16"). Top dia.: 7.1 cm (213/16"). Top dia. with spout: 7.5 cm (215/16"). Bottom dia.: 5.1 cm (2").