Sani-Cloth® Plus Germicidal Disposable Wipes, PDI®
Supplier: Healthcare Products
PDI Q85084 PDI Q89072
15648-996CS 198.51 USD
15648-996 15648-990
Sani-Cloth® Plus Germicidal Disposable Wipes, PDI®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Deep well lid provides ample space to store and access next wipe. Prominent contact time for easy identification and improved compliance with accrediation standards. Categorization of pathogens allows for quick identification of relevant efficacy claims. Safety icon clearly communicates key safety information on both label and lid.
  • EPA-registered, low-level disinfectant
  • Pre-moistened and ready-to-use
  • Cleans and disinfects in one step
  • Bactericidal and virucidal
  • Compatible with a broad range of hard nonporous surfaces
  • Reduces waste and saves time
Certifications: Meets CDC, OSHA, AHE and CMS Tag F441 guidelines.