PVP Iodine Prep Pads, PDI®
Supplier: Healthcare Products
PDI B40600
15648-926BX 30.65 USD
15648-926
PVP Iodine Prep Pads, PDI®
Wipes Cleaning Kits and Pads
These pads are saturated with a 10% USP Povidone-Iodine prep solution to provide proven antisepsis.
They are non-irritating and virtually non-stinging for greater patient comfort.