Woll Drain Dissecting Tray, Shandon, Thermo Scientific
Supplier: Epredia
360
15640-516EA 491.63 USD
15640-516
Woll Drain Dissecting Tray, Shandon, Thermo Scientific
Trays/Dishes/Pots Dissecting Trays
Designed to restrict contamination by directing spillage into a sink
Useful for cleaning glassware, urinalysis, solution preparation, bacterial preparation, and other lab applications. Spout at the end of the tray is 12.7Wx7.6Lcm (5x3") with a 20˚ slope. Adjustable tray slope.