VWR® Blood Collection Tray
Supplier: VWR International
15600-250
15600-250CS 881.95 USD
15600-250
VWR® Blood Collection Tray
Trays/Dishes/Pots Blood Collection Trays
This versatile blood collection tray carries all you need from the lab to your patient, and can be rearranged to meet your specific needs.
  • Sturdy polypropylene plastic construction holds its shape even when fully loaded
  • Steam autoclavable at 121°C (250°F)
  • Detachable handle is 17.8 cm (7") high
Blood collection trays are built of sturdy polypropylene plastic that resists warping even when fully loaded.
Ordering information: Includes two, 40-place test tube racks for tubes up to 16 mm O.D., one 18-place slide holder, a large requisition section, and adjustable partitions to hold needles, syringes, cotton and other materials.